Learn More
Lakeland Industries High Visibility
Supplier:  Lakeland Industries V10AM2023ZL2X
Encompass_Preferred
Lakeland Class 3 FR/AR Static Dissipative Mesh Hi-Vis Vests are designed to be cooler and more comfortable, without sacrificing protection. V10AM2023ZL is certified to NFPA 70E CAT 1 and ANSI Class 3, with non-conductive zipper closure, reinforced lanyard port, and 3M Scotchlite reflective material. Available in sizes MD-5X, these sleeved vests are ideal for work environments where visibility is important, and when flash fire is a concern.
Catalog No. 50-284-1217