Learn More
Lakeland Industries High Visibility
Supplier:  Lakeland Industries V8AM0322VL3X
Encompass_Preferred
Lakeland Class 2 FR Modacrylic Mesh Hi-Vis Vests are designed with FR treated mesh fabric, and reflective material. V8AM0322VL meets ANSI 107-2015 Class 2. Available in sizes MD-5X, these vests are ideal for worker environments where visibility is important, and when flash fire is a concern.
Catalog No. 50-284-0989