Learn More
Lakeland Industries High Visibility
Supplier:  Lakeland Industries V8AM0123ZL4X
Encompass_Preferred
Class 3, FR/AR Modacrylic Mesh Hi-Vis Vest Non-conductive zipper closure, open sides with adjustable sides-snap closure. Built with GlenGuard? Hi-Vis fabric which is patented and inherently flame resistant. Designed to make safety vests cooler and more comfortable without sacrificing protection. The 2" 3M Scotchlite Retroreflectivity Material helps the eye perceive objects in low-light conditions when illuminated by a light source. ANSI 107-2015 Class 3 vest 2" 3M Scotchlite Reflective Material Non-conductive zipper front closure Orange trim 2 front slant pockets 27" length
Catalog No. 50-284-0473