Learn More
Lakeland Industries High Visibility
Supplier:  Lakeland Industries V10AM0123ZL6X
Encompass_Preferred
Class 3, FR/AR Static Dissipative Mesh Hi-Vis Vest with Orange Modacrylic Binding Built with GlenGuard? Hi-Vis Static Dissipative Modacrylic fabric which is patented and inherently flame resistant. Designed to make safety vests cooler and more comfortable without sacrificing protection. The fabric is designed with a 10mm carbon core grid-shaped fiber which works to dissipate a potentially harmful static charge allowing safe work in potentially combustible environments. The 2" 3M Scotchlite Retroreflectivity Material helps the eye perceive objects in low-light conditions when illuminated by a light source. ANSI 107-2015 Class 3 2" 3M Scotchlite Reflective Material Non-conductive zipper front closure Open sides with adjustable snap closure 27" length
Catalog No. 50-284-0484