Learn More
TA Instruments High Volume Sample Pans Lids and Seals, 100 Sets DSC High Volume Sample Pans 100µL, Lids and Seals, Suppress Vaporization to 250°C, Requires High Volume Sample Pan Kit, Operational Use, 100 Pack
Supplier:  TA Instruments 900796.901
Encompass_Preferred
Waters TA Instruments High Volume Sample Pans Lids And Seals contain 100 sets of high-volume DSC sample pans (100 µL), lids, and seals that suppress vaporization to 250°C. Requires High Volume Sample Pan Kit for sealing and operational use.
Catalog No. 50-005-80884