Learn More
Sigma Aldrich Fine Chemicals Biosciences HiLoadR 26/600 SuperdexR 2
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE28989336
Encompass
HiLoad(R) 26/600 Superdex(R) 200 prep grade are prepacked XK columns designed for high-resolution preparative gel filtration chromatography. The column is an excellent choice highly suitable for separating MAbs from dimers and contaminants.Superdex(R) prep grade (pg) is produced by covalent bonding of dextran to highly cross-linked agarose.The separation properties of these media are determined by the dextran component. Steep selectivity curves give excellent resolving power for peptides and proteins in the molecular weight ranges Mr < 10 000 (Superdex(R) 30 pg) Mr 3 000 to 70 000 (Superdex(R) 75 pg) and Mr 10 000 600 000 (Superdex(R) 200 pg). The medium combines high mechanical strength with high hydrophilicity allowing high flow rates and minimal nonspecific interactions.
Catalog No. 50-290-8625
Only null left
Add to Cart
Request a Quote