Learn More
Medchemexpress LLC Himic anhydride | 2746-19-2 | MFCD00630580 | 164.16 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0082935G
Encompass_Preferred
Himic anhydride is a compound related to other acid anhydrides, serving as an important intermediate in the synthesis of pharmaceuticals, pesticides, materials, and resins. This chemical can be utilized in the production of brominated flame retardants and has been noted to elicit IgE-mediated sensitization in the workplace. Research indicates its potential use in the study of asthma.
  • Acts as an important intermediate in various syntheses
  • Used in the production of brominated flame retardants
  • May cause IgE-mediated sensitization
  • Applicable in asthma research
Catalog No. 50-004-46869