Learn More
Electron Microscopy Sciences HINGED PLASTC CONTNRS 21.29ML
SDP
Supplier:  Electron Microscopy Sciences 7261705M
Encompass_Preferred
These one piece hinged plastic round containers are made from Medical-Grade Polypropylene. Perfect in labs settings for dry or wet samples. They are air and moisture resistant. Have non-allergenis animal-free components and autoclavable (sterilizable) in the presence of aqueous solutions. Food packaging safe. Available in a variety of sizes.
Catalog No. 50-241-3081