Learn More
Enzo Life Sciences Hinokitiol (250mg). CAS: 499-44-5
SDP
Supplier:  Enzo Life Sciences BMLFR1100250
Encompass_Preferred
Hinokitiol (β-thujaplicin) is a potent metal chelator that induces differentiation and apoptosis in teratocarcinoma F9 cells and tyrosinase, most likely via metal chelation. Purity: ≥98% (GC). Long Term Storage: Ambient.
Catalog No. 50-201-0831