Learn More
Medchemexpress LLC Hippuric acid-d5 | 53518-98-2 | 99.7% | 184.20 | C9H4D5NO3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW016562S10MG
Encompass_Preferred
Hippuric acid-d5 is the deuterium-labeled analogue of hippuric acid used as a stable isotope internal standard and tracer for quantitative analyses. It is intended for use in NMR, GC-MS, and LC-MS workflows to improve accuracy and reproducibility when measuring hippuric acid in biological or environmental samples.
  • Deuterium-labeled internal standard for quantitative assays.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • High chemical purity for reliable quantification.
  • Stable solid form for convenient storage and handling.
Catalog No. 50-003-00783