Learn More
Medchemexpress LLC Hippuryl-Arg (Bz-Gly-Arg) | 744-46-7 | MFCD00055706 | 99.9% | 335.36 g/mol | C15H21N5O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1316185MG
Encompass_Preferred
Hippuryl-Arg is a synthetic peptide substrate used to detect and quantify enzymes that recognize and cleave peptide bonds containing arginine. Supplied as a white to off-white solid with high purity, it is suitable for biochemical and enzymatic assays where a reliable arginine-specific substrate is required. The compound dissolves in DMSO with heating and ultrasonic assistance and has defined storage conditions to preserve stability.
  • high purity for consistent assay performance.
  • soluble in DMSO with warming for convenient preparation.
  • defined storage stability for long-term powder storage.
  • suitable for fluorescence or UV-based enzyme assays.
  • provides known molecular weight and formula for experiment planning.
Catalog No. 50-002-62576