Learn More
Medchemexpress LLC Hippuryl-His-Leu-OH hydrate (N-Benzoyl-Gly-His-Leu hydrate) | 207386-83-2 | 99.7% | 429.47 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13787550MG
Encompass_Preferred
Hippuryl-His-Leu-OH hydrate (N-Benzoyl-Gly-His-Leu hydrate) is a specific substrate for angiotensin-converting enzyme (ACE) I, used as a molecular tool for detecting ACE activity in in vitro experiments. It is hydrolyzed by ACE through competitive binding, producing hippuric acid, which can be quantified to assess ACE activity or screen for ACE inhibitors. The released His-Leu can also be detected via fluorescence.
  • Specific substrate for angiotensin-converting enzyme (ACE) I.
  • Molecular tool for detecting ACE activity in vitro.
  • Hydrolyzed by ACE to produce hippuric acid.
  • Quantitatively assesses ACE activity.
  • Screens for ACE inhibitors.
  • Released His-Leu detectable via fluorescence.
  • Applicable for hypertension and cardiovascular disease research.
  • Used in studying changes in ACE activity during physiological and pathological processes.
Catalog No. 50-003-50688