Learn More
Sigma Aldrich Fine Chemicals Biosciences HiPrepTM 26 0 Desalting C
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE17508702
Encompass
HiPrep(TM) 26/10 Desalting is a convenient easy-to-use column prepacked 53 mL column with Sephadex(R) G-25 Fine for desalting and buffer exchange.
Catalog No. 50-290-7231