Learn More
Sigma Aldrich Fine Chemicals Biosciences HiPrepTM Sp Fast Flow 16
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE28936544
Encompass
HiPrep(TM) SP FF is a strong cation exchanger prepacked with SP Sepharose(R) Fast Flow ready-to-use for fast preparative separations of proteins and other biomolecules using ion exchange (IEX) chromatography.HiPrep(TM) SP FF are prepacked ready to use columns for ion exchange chromatography. They provide fast preparative separations of proteins and other biomolecules.
Catalog No. 50-290-7191