Learn More
Sciencell Research Laboratories HiPSC-derived Endothelial Cells, Includes 1 vial of HiPSC-EC
Supplier:  Sciencell Research Laboratories 1690
Encompass
Human Endothelial Cells (HiPSC-EC) from ScienCell Research Laboratories are differentiated from a human induced pluripotent stem cell line (HiPSC), which is generated using mRNA reprogramming technology from Human Dermal Fibroblasts (HDF). The monolayer HiPSC efficiently generate mesoderm cells via small-molecule activation of WNT signaling in chemically defined media, and subsequently converted to endothelial cells through endothelial cell specification using vascular endothelial growth factor (VEGF) and fibroblast growth factor (FGF).The derived endothelial cells are characterized by immunofluorescence with antibodies specific to CD31 and vWF. The cell population is highly pure: >80% of cells express CD31 (PECAM-1) & vWF positive. HiPSC-EC are cryopreserved at passage one and delivered frozen. Each vial contains >1 x 106 cells in 1 ml volume. Cells are negative for mycoplasma, bacteria, yeast and fungi.
Catalog No. NC3367419