Learn More
Medchemexpress LLC His-[D-2-ME-Trp]-Ala | 3026595-80-9 | 99.9% | 426.47 | 10 MG
Supplier: Medchemexpress LLC HYP146010MG
His-[D-2-ME-Trp]-Ala is a fragment of the growth hormone hexarelin.
- Sealed storage, away from moisture and light.
- Powder: -80°C for 2 years; -20°C for 1 year.
- In solvent: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture and light).