Learn More
Medchemexpress LLC His-D-beta-Nal-Ala-Trp-D-Phe-Lys-NH2 TFA | 99.6% | 998.06 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP5189A5MG
Encompass_Preferred
His-D-beta-Nal-Ala-Trp-D-Phe-Lys-NH2 TFA is a growth hormone releasing peptide and a metabolite of GHRP-1. GHRP-1 promotes the release of growth hormone (GH) and increases intracellular calcium levels in static monolayer cells of rat pituitary gland, but does not affect cAMP levels.
  • Growth hormone releasing peptide
  • Metabolite of GHRP-1
  • Promotes growth hormone release
  • Increases intracellular calcium levels in rat pituitary gland cells
  • Does not affect cAMP levels
Catalog No. 50-003-96371