Learn More
MONOD BIO INC Anti-Histidine Tag NovoBody™ Duo Binder, Biotin Labeled
Supplier:  MONOD BIO INC MB7976BIO
Encompass_Preferred
NovoBody™ Duo Biotinylated Anti-His TagThe biotinylated NovoBody™ Duo is a bivalent anti-His Tag binder with enhanced affinity over conventional antibodies. Biotinylated via amine chemistry (degree of labeling 2–4).NovoBody™ Binders are single-chain proteins designed computationally to improve on traditional antibodies. They offer enhanced stability and function while eliminating suboptimal traits like post-translational modifications. Suitable for ELISA, flow cytometry, and capture applications.Tag: Twin-Strep-tag | Conjugation: Biotin | Amount: 100 µgLearn more at monod.bio
Catalog No. NC3705681