Learn More
Epigentek Inc Histone H3K9me3 (H3K9 Trimethyl) Polyclonal Antibody (50 ul)
SDP
Supplier:  Epigentek Inc A4036050
Encompass_Preferred
Unconjugated Polyclonal Antibody Raised in Rabbit. Immunogen: Synthetic Peptide of Human TriMethyl-Histone H3-K9. Tested Applications: WB, IHC, IF, IP, ChIP, ChIPseq. Reactivity: Broad Range, Mouse, Rat, Human. Alternative Names: H3K9me3 antibody; H3K9m3 antibody; HIST1H3J; H3/j; H3FJ; Histone H3.1; Histone H3/a; Histone H3/b; Histone H3/c; Histone H3/d; Histone H3/f; Histone H3/h; Histone H3/I; Histone H3/j; Histone H3/k; Histone H3/l; HIST3H3 .
Catalog No. 50-109-5515