Learn More
Abcam Histamine, histamine receptor agonist, 5G
Supplier:  Abcam AB1462415G
Encompass_Preferred
MW 111.1 g/mol. Potent vasodilator. Formed by the decarboxylation of L-histidine. Acts as an agonist for endogenous histamine receptors. Histamine H₁ receptor activation mobilizes Ca²+; H₂ activation stimulates adenylate cyclase activity in neurons. Activates nitric oxide synthase (NOS).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4807