Learn More
Sigma Aldrich Fine Chemicals Biosciences Histamine Quantification A
Supplier:  Sigma Aldrich Fine Chemicals Biosciences MAK4321KT
Encompass
Histamine is a biogenic amine generated from the single decarboxylation of the amino acid histidine. Histamine food poisonings are allergy-like food poisonings caused by the ingestion of spoiled food containing markedly elevated histamine levelsElevated levels of bacterial fermentation can lead to elevated histamine in various food and drink sources such as fish fermented meat milk cheese beer and wine.
Catalog No. 50-290-4261