Learn More
Leica Biosystems HistoCore Water Bath M - compact histology water bath for slide flotation and preparation (Model 140607020C1)
Supplier:  Leica Biosystems 140607020C1
Encompass
The HistoCore Water Bath M is a compact water bath designed for histology laboratories to flatten floating tissue ribbons and assist slide preparation. Intended for in vitro diagnostic use, it can be paired with an optional slide dryer to speed water evaporation and simplify slide handling.
  • Detachable water tray for easy cleaning and replacement.
  • Black anodized tray for improved visibility and contrast.
  • Auxiliary illumination to aid ribbon placement and visibility.
  • OLED touch panel with intuitive user interface for precise control.
  • Dual voltage operation (100-120 V and 220-240 V) and fast heating up to 60 °C.
  • Compact footprint (280 x 280 x 105 mm) and lightweight design for benchtop use.
Catalog No. NC3373748