Learn More
Cayman Chemical HIston H2AC-Term Monclon 100ug
SDP
Supplier:  Cayman Chemical 32181100 UG
Encompass_Preferred
Immunogen: Peptide from the C-terminal region of human H2A • Host: Rabbit • Species Reactivity: (+) Vertebrates • Cross Reactivity: (+) H2A independent of PTMs; (-) H2B, other histone proteins • Applications: ELISA, ICC, multiplex-based assays, WB
Catalog No. 50-294-9211