Learn More
Cayman Chemical HIston H3K27Me3 MoncLNal A
SDP
Supplier:  Cayman Chemical 20719100 UG
Encompass_Preferred
Immunogen: Peptide corresponding to H3K27Me3 • Host: Rabbit • Species Reactivity: (+) Vertebrates • Cross Reactivity: (+) H3K27Me3; (-) Unmodified histone H3K27, H3K27Me1, H3K27Me2, H3K4Me1, H3K4Me2, H3K4Me3, H3K9Me1, H3K9Me2, H3K9Me3, H3K14Me2, H3K18Me1, H3K18Me2, H3K23Me1, H3K23Me2, H3K36Me1, H3K36Me2, H3K36Me3, H3K56Me1, H3K79Me1, H3K79Me2, H3K79Me3 • Applications: ChIP, ELISA, IHC, multiplex-based assays, WB
Catalog No. 50-298-3194