Learn More
Abcam Histone Acetyltransferase Inhibitor II, p300/CBP HAT inhibitor, 10MG
Supplier:  Abcam AB14455610MG
Encompass_Preferred
MW 464.1 Da, Purity >98%. Selective, cell-permeable p300/CBP histone acetyltranserase inhibitor (IC₅₀ = 5 μM). Antitumor agent. Active in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5635