Learn More
CPC Scientific H-Lys-Asp-Thr-Lys-Pro-Thr-Pro-Thr-Lys-Gly-Lys-Ala-Ala-Ser-Ala-Ser-Thr-Thr-Pro-Val-Lys-Lys-Asp-Val-Thr-Pro-Val-Lys-Ala-Asp-Thr-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIST010A
Encompass
This peptide is histone H1 (29-59) derived from Tetrahymena thermophila.ONE-LETTER SEQUENCE: KDTKPTPTKGKAASASTTPVKKDVTPVKADTMOLECULAR FORMULA: C137H238N38O47MOLECULAR WEIGHT:3169.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: Miscellaneous
Catalog No. 50-210-9440