Learn More
CPC Scientific H-Ala-Thr-Lys-Ala-Ala-Arg-Lys-Ser-Ala-Pro-Ala-Thr-Gly-Gly-Val-Lys-Lys-Pro-His-Arg-Tyr-Arg-Pro-Gly-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIST048A
Encompass
This peptide is the fragment of Histone H3 (21-44).ONE-LETTER SEQUENCE: ATKAARKSAPATGGVKKPHRYRPGMOLECULAR FORMULA: C109H185N39O29MOLECULAR WEIGHT:2505.91STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [1373516-71-2], SYNONYMS: H3 (21-44)RESEARCH AREA: Miscellaneous
Catalog No. 50-210-9488