Learn More
Genscript Corporation Histone H3 His Human
Supplier:  Genscript Corporation Z02527
Encompass
Histone H3 is one of the five main histones involved in the structure of chromatin in eukaryotic cells Featuring a main globular domain and a long N-terminal tail H3 is involved with the structure of the nucleosomes of the beads on a string structure Histone proteins are highly post-translationally modified however Histone H3 is the most extensively modified of the five histones Histone H3 is an important protein in the emerging field of epigenetics where its sequence variants and variable modification states are thought to play a role in the dynamic and long term regulation of genes
Catalog No. 50-000-59469