Learn More
Sino Biological Peptide Substrates: Histone H3 Peptide (1-21)
Supplier:  Sino Biological H1258500
Encompass_Preferred
The Histone H3 Peptide (1-21) sequence (ARTKQTARKS-TGGKAPRKQLAGGKKC) is derived from human histone H3 (1-21) and is suitable for use as the substrate for histone methyltransferase (at K4 and K9) and acetyltransferase (at K9 and K14) family assays.
Catalog No. 50-256-9044