Learn More
AnaSpec Histone H4 (1-23) - GGK(Biotin), biotin labeled, amide, 1mg
Supplier:  AnaSpec AS65097
Encompass
Histone H4 (1-23)-GGK(Biotin), biotin labeled, amide[SGRGKGGKGLGKGGAKRHRKVLRGG-K(Biotin)-NH2] ; MW 2828.4; (1 mg) This peptide is of Histone H4 (1-23) biotinylated through a GGK linker on the C-terminus. Provided at 95% peptide purity, this peptide was dissolved in distilled water at 1 mg/ml and re-lyophilized to powder form.
Catalog No. NC0061325