Learn More
Epigentek Inc EpiQuik Histone H4 Modification Multiplex Assay Kit (Colorimetric) new product  (96 Assays)
SDP
Supplier:  Epigentek Inc P310296
Encompass_Preferred
For ELISA-like quantitation of 10 different histone H4 modification patterns on the same plate
Catalog No. NC1526875