Learn More
Sino Biological Peptide Substrates: Histone H4 Peptide (1-21)
Supplier:  Sino Biological H135801
Encompass_Preferred
The Histone H4 Peptide (1-21) sequence (SGRGKGGKGL-GKGGAKRHRKVGGKKC) is derived from human histone H4 (1-21) and is suitable for use as the substrate for histone methyltransferase (at R3) and acetyltransferase (at K5, K8, K12 and K16) family assays.
Catalog No. 50-256-9042