Learn More
Supply Solutions Histopaque-1077, 6x100mL
Supplier:  Supply Solutions SIGMA 107716X100ML
Encompass_Preferred
Sterile-filtered, density: 1.077 g/mL, Histopaque-1077 is a sterile, endotoxin tested solution of polysucrose and sodium diatrizoate, adjusted to a density of 1.077g/mL. This ready-to-use medium facilitates rapid recovery of viable lymphocytes and other mononuclear cells from small volumes of whole blood.
Catalog No. 50-161-3321