Learn More
Nacalai USA Inc HistoVT One (10x, pH 7.0), Antigen Retrieval Solution, 100ml
Supplier:  Nacalai USA Inc 0638076
Encompass
HistoVT One is an antigen retrieval solution for immunohistochemistry and in situ hybridization. This product can unmask antigenic sites without damage to antigen from formalin-fixed, frozen or paraffin-embedded tissue sections.
Catalog No. NC3714945