Learn More
Sigma Aldrich Fine Chemicals Biosciences HiTrapR CaptoTM S Cytiva 1
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE17544123
Encompass
HiTrap(R) Capto(TM) S is a strong cation exchanger prepacked with BioProcess Capto(TM) media for screening and small-scale protein purifications using ion exchange chromatography (IEX).
Catalog No. 50-290-4172