Learn More
Sigma Aldrich Fine Chemicals Biosciences HiTrapR DEAE fast flow Cyt
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE17515401
Encompass
HiTrap(R) DEAE ff is prepacked with DEAE Sepharose fast flow and is a weak anion exchanger for small-scale protein purifications as well as screening of binding and elutions conditions.Q SP DEAE and CM Sepharose(R) fast flow are based on a robust 6% highly cross-linked beaded agarose matrix with good flow properties and high loading capacities. ANX Sepharose(R) 4 fast flow (high sub) is based on 4% highly cross-linked beaded agarose. This results in a medium with higher porosity which is particularly useful for the purification of high molecular mass proteins.The active end of the charged group is the same for DEAE Sepharose fast flow and ANX Sepharose(R) fast flow (high sub) the difference is the length of the carbon chain of the charged group. DEAE Sepharose fast flow has a diethylaminoethyl-group bound to the agarose whilst ANX Sepharose(R) 4 fast flow (high sub) has a diethylaminopropyl group attached.
Catalog No. 50-290-4087