Learn More
Sigma Aldrich Fine Chemicals Biosciences HiTrapR IMAC Fast Flow Cyt
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE17092104
Encompass
HiTrap(R) IMAC FF is prepacked with IMAC Sepharose(R) 6 Fast Flow. The medium is charged with the metalion of your choice for Immobilized Metal ion Affinity Chromatography (IMAC) and subsequent purification of polyhistidine tagged proteins.
Catalog No. 50-290-7645