Learn More
Sigma Aldrich Fine Chemicals Biosciences HiTrapR Phenyl High Perfor
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE17519501
Encompass
HiTrap(R) Phenyl HP are prepacked Phenyl Sepharose(R) High Performance columns for high-reSOLUTIONon hydrophobic interaction chromatography (HIC). The columns are used for small-scale protein purification as well as for screening of binding and elutions conditions.
Catalog No. 50-290-7823