Learn More
Sigma Aldrich Fine Chemicals Biosciences HiTrapR Protein G High Per
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE17040403
Encompass
HiTrap(R) Protein G HP columns are prepacked with Protein G Sepharose(R) High Performance for fast easy separations of IgG from different species and subclasses.
Catalog No. 50-290-4323