Learn More
CPC Scientific H-Asp-Thr-Gly-Ser-Ser-Ser-Lys-Val-Ser-Gln-Asn-Tyr-Pro-Ile-Val-Gln-Asn-Ala-Gln-Gly-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific HIV013A
Encompass
ONE-LETTER SEQUENCE: DTGSSSKVSQNYPIVQNAQGMOLECULAR FORMULA: C86H138N26O34MOLECULAR WEIGHT:2080.17STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [1815618-20-2], RESEARCH AREA: Antimicrobial
Catalog No. 50-210-9553