Learn More
Medchemexpress LLC HIV-1 inhibitor-8 | 2826996-78-3 | MFCD34567332 | 98.0% | 439.53 | C25H21N5OS | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1322915MG
Encompass_Preferred
HIV-1 inhibitor-8 is an orally active, low-toxicity non-nucleoside reverse transcriptase inhibitor used in research to evaluate antiviral activity against HIV-1 strains.
  • Orally active non-nucleoside reverse transcriptase inhibitor for research use.
  • Potent antiviral activity (EC50 = 4.44-54.5 nM) against various HIV-1 strains.
  • Inhibits wild-type HIV-1 reverse transcriptase (IC50 = 0.081 μM).
  • High chemical purity (98.0%).
  • Molecular formula C25H21N5OS and molecular weight 439.53 g/mol.
  • Supplied as a 5 mg research quantity.
Catalog No. 50-002-63145