Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HIV-1 integrase inhibitor 8 | 1568-80-5 | 99.9% | 308.41 g/mol | C21H24O2 | 100 MG
SDP

Supplier:  Medchemexpress LLC HY107485100MG

Encompass_Preferred

HIV-1 integrase inhibitor 8 is a small-molecule research compound that inhibits HIV-1 integrase. Supplied as a powdered solid, it is intended for laboratory research use only and not for human or veterinary applications.

  • Intended for in vitro research use only.
  • Powdered solid appearance.
  • Molecular formula C21H24O2; molecular weight 308.41 g/mol.
  • Purity 99.9%.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Available in 50 mg and 100 mg quantities.
  • CAS number 1568-80-5.

Catalog No. 50-002-39912


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.