Learn More
ProteinSimple HIV-1 p24 2nd Gen Cartridge, Assay Cartridge, 72 samples, 11.7-112,000 pg/mL detection range, less than 90 min reaction time
Supplier:  ProteinSimple SPCKBCS009216
Encompass
HIV-1 p24 2nd Gen Cartridge, Assay Cartridge, 72 samples, 11.7-112,000 pg/mL detection range, less than 90 min reaction time
Catalog No. NC2601629