Learn More
XpressBio HIV-1 p24 ELISA Kit, 96 wells
SDP
Supplier:  XpressBio XB1000
Encompass_Preferred
ELISA plate wells coated with anti HIV-1 p24 murine monoclonal antibody. The assay is used to monitor HIV-1 in cell culture or to determine the viral titer of lentiviral samples.
Catalog No. NC0904142