Learn More
Creative Biomart Recombinant HIV-1 p66 pol Antigen
Supplier:  Creative Biomart HIV284
Encompass_Preferred
HIV-1 p66 Recombinant-is a 71 kDa protein derived from pol gene. The HIV-1 p66 is glycosylated with N-linked sugars and produced using baculovirus vectors in insect cells.
Catalog No. 50-208-4662