Learn More
Sino Biological HIV Gag-p24 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 11695MM1520
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant HIV-1 p24 (group M, subtype B, strain 92418) ( Catalog#11695-V08E; ACI05538.1; Pro133-Leu363). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-2015