Learn More
Medchemexpress LLC Hjc0152 | 1420290-99-8 | MFCD29770804 | 99.8% | 406.65 g/mol | C15H14Cl3N3O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10060210MG
Encompass_Preferred
HJC0152 is a small-molecule inhibitor of signal transducer and activator of transcription 3 (STAT3) for research use; it inhibits STAT3 signaling, decreases STAT3 promoter activity, reduces total STAT3, induces cleaved caspase-3, down-regulates cyclin D1, and shows antitumor activity in mouse xenograft models with no significant toxicity at tested doses.
  • Inhibits STAT3 signaling and promoter activity in vitro.
  • Reduces cell proliferation and induces apoptosis at low micromolar concentrations.
  • Demonstrated tumor growth inhibition in mouse xenograft models without notable toxicity.
  • High purity suitable for research applications (~99.8%).
  • Available in small quantities for in vitro and in vivo studies.
Catalog No. 50-002-14721