Learn More
Medchemexpress LLC HJC0152 hydrochloride | 1420290-99-8 | MFCD29770804 | 99.8% | 406.65 g/mol | C15H14Cl3N3O4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY100602100MG
Encompass_Preferred
HJC0152 is a small-molecule inhibitor of signal transducer and activator of transcription 3 (STAT3) intended for research use. It is used in cellular and biochemical studies to probe STAT3 signaling and evaluate antitumor effects in preclinical experiments.
  • Potent inhibitor of STAT3 signaling in cellular assays.
  • Demonstrated to inhibit cell cycle progression and induce apoptosis.
  • High reported purity (99.75%).
  • Suitable for biochemical and cell-based research applications.
  • Available in multiple milligram pack sizes for flexible dosing.
  • Solid form; store sealed at 4°C and protect from moisture.
Catalog No. 50-002-14584