Learn More
TARGETMOL CHEMICALS INC HJC0152 HYDROCHLORIDE 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T423425MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. HJC0152 hydrochloride (HJC0152) is a signal transducer and activator of transcription 3 (STAT3) inhibitor. purity: 98%
Catalog No. 50-257-9922