Learn More
Medchemexpress LLC HY-103443 10mg Medchemexpress, HKI-357 CAS:848133-17-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10344310MG
Encompass_Preferred
Medchemexpress, HY-103443 10mg HKI-357 CAS:848133-17-5 HKI-357 is an irreversible dual inhibitor of EGFR and ERBB2 with IC50s of 34 nM and 33 nM, respectively. HKI-357 suppresses EGFR autophosphorylation (at Y1068), and AKT and MAPK phosphorylation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8107